Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Bay 11-7085, Inhibitor, Greater Than 99% Purity, Solid, 10 mg

Catalog No. 502706519
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6519 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6519 Supplier Abcam Supplier No. AB14157410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Irreversibly inhibits TNF-α-induced IxBα phosphorylation to reduce NF-κB and adhesion molecules.

  • Molecular Weight: 249.33 Da
  • Purity: >99%
  • Form: Solid
  • Mechanism: Irreversibly inhibits TNF-α-induced IxBα phosphorylation
  • Activity: Reduces NF-κB and adhesion molecules (IC50 = 5-10 μM)
  • Applications: Anti-inflammatory, anti cancer and neuroprotective agent in vivo
  • CAS Number: 196309-76-9

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule IκBα
Shipping Condition Ambient
Alternate Part Number AB141574
Content And Storage Store at ambient temperature.
Form Solid
Quantity 10 mg
For Use With (Application) Anti-inflammatory, Anti-cancer, Neuroprotection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.