Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam BAY 11-7085, NF-kB Inhibitor, >99% Purity, Solid, 50 mg

Catalog No. 502706518
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6518 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6518 Supplier Abcam Supplier No. AB14157450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent and irreversible inhibitor of TNF-α-induced IκBα phosphorylation with anti-inflammatory and neuroprotective properties.

  • Molecular Weight: 249.33 Da
  • Purity: >99%
  • Mechanism of Action: Irreversibly inhibits TNF-α-induced IκBα phosphorylation
  • Potency: Reduces NF-κB and adhesion molecules (IC50 = 5 to 10 μM)
  • Applications: Anti-inflammatory, anticancer and neuroprotective agent in vivo

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule IκBα
Shipping Condition Ambient
Buffer 100% BAY 11-7085
Content And Storage Store at ambient temperature.
Form Solid
Formulation 100% BAY 11-7085
Quantity 50 mg
For Use With (Application) Anti-inflammatory, anti cancer, neuroprotection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.