Learn More
TARGETMOL CHEMICALS INC BAY 2965501 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T73053
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. BAY 2965501 is a potent and selective diacylglycerol kinase zeta (DGK(zeta)) inhibitor that induces pERK activation.BAY 2965501 can be used in the study of cancer. purity: 99%
Catalog No. 50-257-9224
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.