Learn More
TARGETMOL CHEMICALS INC BAY 60-6583 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1450610MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BAY 60-6583 is a potent high-affinity adenosine A2B receptor agonist (EC50 = 3 nM) with affinity for A2B receptors that exceeds that for A1 A2A and A3 receptors.BAY 60-6583 is cardioprotective in a model of myocardial ischemia.The Ki values for BAY 60-6583 binding to mouse rabbit and dog A2BAR were 750 nM 340 nM and 330 nM respectively. The Ki values of BAY 60-6583 binding to A2BAR in mice rabbits and dogs were 750 nM 340 nM and 330 nM respectively. purity: 99%
Catalog No. 50-257-7382
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.