Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ BAY 8002

Catalog No. p-7166801
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-171-0 10 mg
68-175-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 68-171-0 Supplier Tocris™ Supplier No. 6817/10
This item is not returnable. View return policy

Potent dual MCT1/2 inhibitor; orally bioavailable

BAY 8002 is a potent dual MCT1/2 inhibitor (IC50 values are 3, 8, and 12 nM at rat (C6), human (DLD-1) and mouse (4T1) MCT1 respectively; IC50 values of human MCT1, MCT2, and MCT4 isoforms recombinantly expressed in X. laevis oocytes are 1, 5 and >500 nM, respectively); suppresses bidirectional lactate transport. Inhibits cellular SNARF-5 fluorescence change (IC50 = 85 nM) in MCT1-expressing DLD-1 cells. Arrests growth and increases intratumor lactate levels of Raji tumor xenografts in mice. Orally bioavailable.

Specifications

Quantity 10 mg
Molecular Formula C20H14ClNO5S
Formula Weight 415.85
Product Type Inhibitor
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.