Learn More
Selleck Chemical LLC BAY-876 5mg 1799753-84-6
SDP
Supplier:  Selleck Chemical LLC S84525MG
Encompass_Preferred
BAY-876 is a potent and selective GLUT1 inhibitor (IC50=0.002 ?M) with a selectivity factor of >100 against GLUT2, GLUT3, and GLUT4. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6069
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.