Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Lauryl Sulfate Broth

Catalog No. B11338 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
11.3 kg
Packaging Type:
Bottle
Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
B11338 500 g Bottle
B4394369 11.3 kg Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B11338 Supplier BD Supplier No. 211338
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For detection of coliform bacteria

Certifications

AOAC, BAM, CCAM, COMPF, EPA, ISO, SMD, SMWW

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, CCAM, COMPF, EPA, ISO, SMD, SMWW
Content And Storage Refrigerate
Form Dehydrated
Media Type Lauryl Sulfate Broth
Product Type Nutrient Broth
Formulation Pancreatic digest of Casein 20g/L, Lactose 5g/L, Dipotassium Phosphate 2.75g/L, Monopotassium Phosphate 2.75g/L, Sodium Chloride 5g/L, Sodium Lauryl Sulfate 0.1g/L
For Use With (Application) Detection of Coliform bacteria
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.