Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Sabouraud Dextrose Agar

Catalog No. B11585 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
2 kg
Packaging Type:
Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
B11585 2 kg Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B11585 Supplier BD Supplier No. 211585

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For cultivation of yeasts, molds and aciduric microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BAM, CCAM, CMPH2, COMPF, EP, JP, MCM9, USP
Content And Storage Refrigerate
Form Dehydrated
Media Type Sabouraud Dextrose Agar
Product Type Agar
Formulation Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 40g/L, Agar 15g/L
For Use With (Application) Cultivation of Yeasts, molds and aciduric microorganisms
Packaging Type Pail
Quantity 2 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.