Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Soy Agar with Lecithin and Polysorbate 80

Catalog No. B12263 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
5 lb.
Packaging Type:
Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
B12263 5 lb. Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B12263 Supplier BD Supplier No. 212263
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms

Detection of microorganisms on surfaces sanitized with quaternary ammonium compounds

Certifications

CE, CCAM

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CCAM
Form Dehydrated
Media Type Soy Agar with Lecithin and Polysorbate 80
Product Type Nutrient Agar
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Lecithin 0.7g/mL, Polysorbate 80 5g/L, Agar 15g/L
For Use With (Application) Detection and enumeration of microorganisms present on surfaces of sanitary importance
Packaging Type Pail
Quantity 5 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.