Learn More
BD BBL™ Gram Stain Kits
Catalog No. B12524 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Product Type:
Gram Stain Kit (Stabilized)
Gram Stain Kit (Unstabilized)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
B12524 Gram Stain Kit (Unstabilized)
B12539 Gram Stain Kit (Stabilized)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12524 Supplier BD Supplier No. 212524
Used to stain microorganisms from cultures or specimens by the differential Gram method
  • Complete stain kits reduce the mess associated with Gram staining procedures
  • Each break-resistant solution bottle features a hinged spot cap that makes regulating the flow easy and helps prevent spots and spills Color-coded labels allow quick identification of reagents
  • Stabilized Iodine Solution requires no mixing and has a long shelf life
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Certifications/Compliance CE
Includes 250mL gram crystal violet, 250mL iodine, 250mL gram decolorizer and 250mL gram safranin
For Use With (Application) Used to stain microorganisms from cultures or specimens by the differential gram method
Stain Type Gram Stain
Product Type Gram Stain Kit (Unstabilized)