Learn More
BD BBL™ Prepared Plated Media: CDC Anaerobe 5% Sheep Blood Agar
Shop All BD Diagnostic Systems ProductsDescription
It also supports the growth of most aerobic, facultatively anaerobic and microaerophilic bacteria if incubated appropriately.
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Light Beige |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | CDC anaerobe 5% Sheep Blood Agar |
| Formulation | Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 20g/L, Yeast extract 5g/L, Hemin 0.005g/L, Vitamin K1 0.01g/L, L-Cystine 0.4g/L, Sheep Blood, defibrinated 5% |
| For Use With (Application) | Isolation and Cultivation of obligate anaerobes from clinical specimens. |
| Packaging Type | Plate |
| Quantity | 20/Pk. |
| Weight (Metric) | 950 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.