Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Columbia CNA Agar with 5% Sheep Blood

Catalog No. L21352 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L21352 20/Pk.
L21353 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. L21352 Supplier BD Supplier No. 221352
This item is not returnable. View return policy

Selective and differential media used for the isolation and differentiation of gram-positive microorganisms from clinical and nonclinical materials

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Red
Content And Storage 2°C to 8°C
Country of Origin United States
Additive Colistin, Nalidixic Acid
Detects Gram-positive Microorganisms
Form Liquid
Format Plate
Manufacturing Origin United States
Media Type Columbia Colistin-Naladixic Acid Agar with 5% Sheep Blood
Product Type Prepared Microbiology Plate
Sterility Sterile
Target Organism Class Gram-positive Organisms
Test Type Microbial Differentiation
Formulation Pancreatic digest of Casein 12g/L, Peptic digest of Animal Tissue 5g/L, Yeast extract 3.5g/L, Beef extract 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Colistin 10mg/L, Nalidixic Acid 10mg/L, Sheep Blood, defibrinated 5%
For Use With (Application) Gram positive microorganisms
Incubation Conditions 35 ±2°C, Aerobic
Incubation Time 24 to 48 hr.
Packaging Type Plate
Quantity 20/Pk.
Weight (Metric) 1300 g
Protein Source Casein, Animal Tissue, Yeast, Beef, Sheep Blood
Manufacturing Quality cGMP, FDA-inspected, ISO 9001 Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.