Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Sabouraud Dextrose Agar (Deep Fill)

Catalog No. B21180X Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21180X 20/Pk.
B21278X 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B21180X Supplier BD Supplier No. 221180
This item is not returnable. View return policy

Used in qualitative procedures for cultivation of dermatophytes

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates
  • Deep-filled plates reduce effects of drying during prolonged incubation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance USP, ISO/DIS, 11014
Color Light Beige
Form Solid
Manufacturing Origin United States
Media Type Sabouraud Dextrose Agar (deep fill)
Formulation Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 40g/L, Agar 15g/L
For Use With (Application) Qualitative procedures for cultivation of Dermatophytes, Candida albicans
Packaging Type Plate
Quantity 20/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.