Learn More
BD BBL™ Prepared Plated Media: Trypticase™ Soy Agar (TSA II™) with Sheep Blood
Shop All BD Diagnostic Systems ProductsDescription
- Ready for immediate use
- Available with 5% and 10% sheep blood
- Deep-fill labelled plates contain additional volume of medium to reduce effects of drying during prolonged incubation
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Red |
| Content And Storage | 2°C to 8°C |
| Country of Origin | United States |
| Form | Prepared Medium |
| Format | Plate |
| Manufacturing Origin | United States |
| Media Type | TSA II with 5% Sheep Blood |
| Product Type | Agar Plate |
| Target Organism Class | Fastidious Organisms |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.