Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Sugar Free Agar

Catalog No. V211672 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V211672 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. V211672 Supplier BD Supplier No. 211672
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the detection and enumeration of organisms in butter and other processed dairy products.

  • Country of Origin: USA
  • Shelf life: 1460 days
TRUSTED_SUSTAINABILITY

Specifications

Form Dehydrated
Manufacturing Origin United States
Product Type Agar
For Use With (Application) Detection and enumeration of organisms in butter and other processed dairy products
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.