Learn More
BBS, MP Biomedicals™
Catalog No. MP116540407 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
200 μL
Catalog No. Quantity
MP116540407 200 μL
1 options
Catalog No. MP116540407 Supplier MP Biomedicals, Inc Supplier No. 116540407
Serves as a general purpose loading dye. MP Biomedicals BBS can be used with neutral DNA or RNA agarose gels.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 200 μL