Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BBS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144794
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144794 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144794 Supplier Invitrogen™ Supplier No. PA5144794
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FIHNPHTRNQ HVSASRVFQS PLESDVSLLN INQAVSCLTA GVLNPELGYD ALLVGTQTNL LAYDVYNNSD LFYREVADGA NAIVLGTLGD ISSPLAIIGG Antigen sequence identity: Mouse 89%; Rat 88%.

This gene is a member of the Bardet-Biedl syndrome (BBS) gene family. Bardet-Biedl syndrome is an autosomal recessive disorder characterized by severe pigmentary retinopathy, obesity, polydactyly, renal malformation and mental retardation. The proteins encoded by BBS gene family members are structurally diverse and the similar phenotypes exhibited by mutations in BBS gene family members is likely due to their shared roles in cilia formation and function. Many BBS proteins localize to the basal bodies, ciliary axonemes, and pericentriolar regions of cells. BBS proteins may also be involved in intracellular trafficking via microtubule-related transport. The protein encoded by this gene forms a multiprotein BBSome complex with six other BBS proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BBS2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BBS2
Gene Accession No. Q9BXC9
Gene Alias 2410125H22Rik; AI447581; Bardet-Biedl syndrome 2; Bardet-Biedl syndrome 2 (human); Bardet-Biedl syndrome 2 homolog; Bardet-Biedl syndrome 2 protein; Bardet-Biedl syndrome 2 protein homolog; BBS; bbs2; bbs2 protein; fb80a05; MGC20703; RP74; Unknown (protein for MGC:134372); wu:fb80a05
Gene Symbols BBS2
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 41-140. Recombinant protein control fragment (Product #RP-109922).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 583
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.