Learn More
Siemens Healthineers BC Thrombin Reagent
Catalog No. 23044705 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
10 x 5 mL
Catalog No. Quantity
23-044-705 10 x 5 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-705 Supplier Siemens Healthineers Supplier No. 10446636
Add to Cart
Add to Cart
This item is not returnable. View return policy
Quantitative determination of thrombin time in citrated human plasma
BC Thrombin reagent is used for the determination of thrombin time. It is suitable for monitoring of fibrinolytic therapy, screening for disorders of fibrin formation, in suspected cases of severe fibrinogen deficiency states, and for differentiation between heparin-induced prolongation of the thrombin time and disorders of fibrinogen formation. Thrombin time is found to be prolonged not only due to disorders in fibrin polymerization, but also due to the presence of heparin. Differentiation can be achieved using Batroxobin reagent.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Thrombin/Batroxobin Time testing
Quantity 10 x 5 mL