Learn More
Siemens Healthineers BC von Willebrand Reagent
Catalog No. 23044754 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
4 x 5 mL
Catalog No. Quantity
23-044-754 4 x 5 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-754 Supplier Siemens Healthineers Supplier No. 10714565
Add to Cart
Add to Cart
This item is not returnable. View return policy
Determination of ristocetin co-factor activity of von Willebrand factor in human plasma through platelet agglutination.
The BC von Willebrand Reagent provides a simple, rapid, and automated procedure for the determination of the ristocetin cofactor activity of von Willebrand factor. Stabilized platelets are agglutinated in the presence of von Willebrand factor and the antibiotic ristocetin A.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) von Willebrand Factor Testing
Quantity 4 x 5 mL