Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam BCA Disodium Salt, Protein Concentration Determination Reagent, Powder, 5 g

Catalog No. 502704711
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4711 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4711 Supplier Abcam Supplier No. AB1463315G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cu(I)-specific colorimetric reagent with >98% purity, used for accurate protein concentration determination.

  • Molecular weight: 388.3 g/mol
  • Purity: >98%
  • Form: Powder
  • Storage buffer: 100% Bicinchoninic acid
  • Mechanism: Cu(I)-specific colorimetric reagent used for measuring protein concentration; proteins reduce alkaline Cu(II) to Cu(I), which complexes with BCA to form a purple complex
  • Detection: Absorbance at 562 nm is directly proportional to protein concentration

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule Protein
Substrate Cu(I)
Shipping Condition Ambient
Buffer 100% bicinchoninic acid
Content And Storage Store at ambient temperature.
Form Powder
Detection Method Colorimetric
Formulation 100% Bicinchoninic acid
Quantity 5 g
For Use With (Application) Protein concentration determination
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.