Learn More
Abcam BCA Disodium Salt, Protein Concentration Reagent, Powder, 25 g
Description
- Molecular weight: 388.3 g/mol
- Purity: >98%
- Form: Powder
- Composition: 100% Bicinchoninic acid
- Mechanism: Cu(I)-specific colorimetric reagent for protein concentration determination
- Detection: Absorbance at 562 nm
- Storage: Ambient
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Protein |
| Detection Location | In-Solution Detection |
| Shipping Condition | Ambient |
| Buffer | 100% Bicinchoninic acid |
| Content And Storage | Store at ambient temperature. |
| Detection Method | Colorimetric |
| Form | Powder |
| Formulation | 100% Bicinchoninic acid |
| Quantity | 25 g |
| For Use With (Application) | Protein concentration determination |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.