Learn More
Specifications
Specifications
| Assay Range | 0.0165 to 1 mg/mL |
| Assay Sensitivity | 0.0165 mg/mL |
| Product Type | Assay Kit (Colorimetric) |
| Sample Type | Serum, Plasma, Cell Culture Supernatant, Tissue, Cells |
| For Use With (Application) | Functional |
| Quantity | 96 Tests |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Target | Non-species specific |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
