Learn More
Cayman Chemical ANTIBODY LY83583 50mg
Supplier: Cayman Chemical 7023050 MG
A diterpene ester with anticancer activity inhibits the growth of K562 HL-60 KT-1 MCF-7/adr A549 and NIH3T3 cells at 12.5 UM induces cell cycle arrest at the G2/M phase apoptosis and necrosis in K562 cells from 31.25-500 nM
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.