Learn More
Apexbio Technology LLC BCATc Inhibitor 2 406191-34-2 5mg
SDP
Supplier:  Apexbio Technology LLC C34635
Encompass_Preferred
BCATc Inhibitor 2 is a small-molecule inhibitor targeting cytosolic branched-chain amino acid transaminase (BCATc) It is designed to inhibit BCATc thereby modulating neuronal glutamate synthesis associated with synaptic excitation and glutamate homeostasis BCATc Inhibitor 2 exerts its biological activity primarily through selective inhibition of cytosolic BCATc affecting calcium influx and glutamatergic signaling in neuronal models Based on these pharmacological properties BCATc Inhibitor 2 holds research potential in studies of neurological and neurodegenerative disorders related to dysfunctional glutamatergic pathways
Catalog No. 50-224-0067