Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ BCC (Body Centered Cube) Molecular Model Kit
SCI_ED

Catalog No. S139061 Shop All Walter Products Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139061 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139061 Supplier Walter Products™ Supplier No. MOLE1013
Add to Cart
Add to Cart

Molecular model kit for the visual representation of cesium chloride.

Cesium chloride is a chemical compound that is used in alternative cancer treatment. This molecular model does a wonderful job of displaying its body centered cube (BCC) molecular structure.

  • Made of durable and colorful plastic
  • Molecular geometry and bond angles can be visibly seen in the structure, making it great for studying
  • Excellent molecular model kit for students studying chemistry in middle school or university
  • These unique molecular structures are great for visual display
  • Ideal for learning about inorganic chemistry

Specifications

For Use With (Application) Inorganic Chemistry
Grade 6+
Material Plastic
Product Type Molecular Model
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.