Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam BCECF-AM, Intracellular Ratiometric pH Indicator, Solid, 1 mg

Catalog No. 502706114
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6114 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6114 Supplier Abcam Supplier No. AB1434631MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Intracellular ratiometric pH indicator that converts to fluorescent BCECF via intracellular esterases for pH measurements between 4 and 9.

  • Application: Intracellular ratiometric pH indicator
  • Chemical properties: Acetoxymethyl ester of BCECF; converts to fluorescent BCECF via intracellular esterases
  • Molecular weight: 556.48 g/mol
  • Purity: >98%
  • Fluorescence characteristics: Excitation at 500 nm (high pH) and 450 nm (low pH); emission at 531 nm
  • pH detection range: 4 to 9

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Form Solid
Target Molecule H+
Substrate Intracellular esterases
Shipping Condition Ambient
Detection Method Fluorescence
Detection Location In-Cell Detection
Content And Storage Store at -20°C.
Buffer 100% 2',7'-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein acetoxymethyl ester
Formulation 100% 2',7'-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein acetoxymethyl ester
Quantity 1 mg
For Use With (Application) Intracellular pH measurement
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.