Learn More
Perkin Elmer US LLC BCECF Excitation Filter Set, 495/440 nm
Supplier:  Perkin Elmer US LLC L2250022
Encompass_Preferred
If using FURA 2 and BCECF, which are both excitation dyes, one drive unit (P/N L2250146) is required to fit on both pairs of filters. Both dyes can be monitored in the same experiment. If using INDO-1 and BCECF, two drive units are necessary- one for excitation and one for emission.
Catalog No. 50-211-2671
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.