Learn More
Bioassay Systems QuantiChrom™ BCG Albumin Assay Kit. For quantitative determination of albumin.
SDP
Catalog No. 50489208 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of albumin and evaluation of drug effects on albumin metabolism. Key Features: Sensitive and accurate. Use as little as 5 µL samples. Detection range 0.01 - 5 g/dL (1.5 - 750 µM) albumin in 96-well plate assay. Simple and high-throughput. The procedure involves addition of a single working reagent and incubation for 5 min. Can be readily automated as a high-throughput assay in 96-well plates for thousands of samples per day. Improved reagent stability and versatility. The optimized formulation has greatly enhanced reagent and signal stability. Cuvet or 96-well plate assay. No interference in biological samples. No pretreatments are needed. Assays can be directly performed on raw biological samples i.e., in the presence of lipid and protein. Method: OD620nm (BCG). Samples: Serum, plasma, urine, biological preparations. Species: All. Procedure: Assay takes 5 min. Kit size: 250 tests. Detection limit: 0.01 g/dL.
Catalog No. 50-489-208 Supplier Bioassay Systems Supplier No. DIAG250
For quantitative determination of albumin and evaluation of drug effects on albumin metabolism. Key Features: Sensitive and accurate. Use as little as 5 µL samples. Detection range 0.01 - 5 g/dL (1.5 - 750 µM) albumin in 96-well plate assay. Simple and high-throughput. The procedure involves addition of a single working reagent and incubation for 5 min. Can be readily automated as a high-throughput assay in 96-well plates for thousands of samples per day. Improved reagent stability and versatility. The optimized formulation has greatly enhanced reagent and signal stability. Cuvet or 96-well plate assay. No interference in biological samples. No pretreatments are needed. Assays can be directly performed on raw biological samples i.e., in the presence of lipid and protein. Method: OD620nm (BCG). Samples: Serum, plasma, urine, biological preparations. Species: All. Procedure: Assay takes 5 min. Kit size: 250 tests. Detection limit: 0.01 g/dL.