Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bcl-2 Monoclonal Antibody (Bcl-2-100), Alexa Fluor™ Plus 647

Catalog No. 756880094
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-688-0094 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 75-688-0094 Supplier Invitrogen™ Supplier No. 756880094
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: This Bcl-2-100 monoclonal antibody reacts to human Bcl-2 (B-cell Lymphoma 2), the founding member of a family of pro-apoptotic proteins which includes Bcl-XL, Bcl-w, and Mcl-1. Bcl-2 plays a role in protecting against apoptosis-inducing insults such as dexamethasone and staurosporine, as well as irradiation and cytokine withdrawal. The mechanism is thought to include regulation of mitochondrial membrane permeability, resulting in sequestration of caspase proforms, or by preventing the release of apoptosis-mediating mitochondrial factors, such as cytochrome c and AIF (apoptosis-inducing factor), into the cytoplasm. Bcl-2 is localized to the cytoplasmic side of the mitochondrial membrane, the nuclear envelope, and the endoplasmic reticulum. Interestingly Bcl-2 does not play a role in preventing cell death of autoreactive T cells in the thymus or Fas-mediated apoptosis. Two isoforms, termed alpha and beta of Bcl-2 have been identified and are a result of alternative splicing at the C-terminus. Applications Reported: This Bcl-2-100 antibody has been reported for use in immunoblotting (WB), immunohistochemistry, immunocytochemistry and flow cytometry. Applications Tested: This Bcl-2-100 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded tissue using high pH antigen retrieval and can be used at 2 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Using conjugate s...

BCL-2 is a key regulator of apoptosis that functions to either inhibit or promote cell death. The BCL-2 family members are also characterized by dimerizing to further modulate apoptosis. Bag1, for example, has been found to form a heterodimer with BCL-2 resulting in the enhancement of the anti-apoptotic effect of BCL-2. Bax and Bak have been shown to play a critical role in cytochrome c release from mitochondria and thus initiate apoptosis. Bax exerts a pro-apoptotic rather than an anti-apoptotic effect on cells. Constitutive expression of BCL2, such as in the case of translocation of BCL2 to Ig heavy chain locus, is thought to be the cause of follicular lymphoma. In most follicular lymphomas, neoplastic germinal centers express high levels of BCL-2 alpha protein, whereas the normal or hyperplastic germinal centers are negative. Two transcript variants of BCL-2, produced by alternate splicing, differ in their C-terminal ends. The overexpression of BCL-2 has been linked to human cancers such as B-cell lymphoma and prostate cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-2
Applications Immunohistochemistry (Paraffin), Multiplex Immunohistochemistry
Classification Monoclonal
Clone Bcl-2-100
Concentration 0.2 mg/mL
Conjugate Alexa Fluor Plus 647
Formulation PBS with 0.5% BSA, 10% proprietary stabilizer and 0.05% sodium azide; pH 7.2
Gene Bcl2
Gene Accession No. P10415
Gene Alias apoptosis regulator Bcl-2; AW986256; B cell leukemia/lymphoma 2; B cell lymphoma 2 associated oncogene; B-cell CLL/lymphoma 2; B-cell leukemia/lymphoma 2; Bcl 2; BCL2; bcl-2; BCL2 apoptosis regulator; Bcl-2 protein; BCL2, apoptosis regulator; Bcl2-like protein; C430015F12Rik; D630044D05Rik; D830018M01Rik; OTTHUMP00000163680; PPP1R50; protein phosphatase 1, regulatory subunit 50
Gene Symbols Bcl2
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues 41-54 of the human Bcl-2 protein. This sequence is not shared by the mouse Bcl-2 protein.
Purification Method Affinity chromatography
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 596
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.