Learn More
Description
Bcl-xL Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | Bcl-xL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 7B2.5 |
| Concentration | 0.1 mg/mL |
| Conjugate | PE |
| Formulation | PBS with proprietary stabilizer and <0.1% sodium azide; pH 7.5 |
| Gene | BCL2L1 |
| Gene Accession No. | P53563, Q07817, Q64373 |
| Gene Alias | anti-apoptosis regulatory protein; apoptosis regulator Bcl-X; B cell lymphoma 2 like; B cell lymphoma like X; B-cell leukemia/lymphoma x; Bcl(X)L; BCL2 like 1; Bcl2l; BCL2L1; bcl2-L-1; BCL2-like 1; Bcl-2-like protein 1; BCLX; bcl-x; BCLXL; Bcl-XL; BCL-XL/S; BCLXS; bcl-xS; Blc2l; DKFZp781P2092; EGK_02534; OTTHUMP00000030550; OTTHUMP00000030552; OTTHUMP00000030555; OTTHUMP00000030556; OTTHUMP00000030557; PPP1R52; protein phosphatase 1, regulatory subunit 52; RP5-857M17.3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
