Learn More
Diagnostic Biosystems BCL6 Bcl-6 (PG-B6)
Supplier:  Diagnostic Biosystems MOB58701
Encompass_Preferred
The BCL6 gene encodes a 706 amino acid nuclear protein of the Kruppel-type zinc finger protein. It is rearranged in about 30% of diffuse large B-cell lymphomas, and is expressed predominantly in normal germinal centre B cells and related lymphomas. The antibody is a useful aid for classification of follicular lymphomas, diffuse large B-cell lymphomas, Burkitt's lymphomas, and nodular, lymphocyte-predominance Hodgkin's lymphoma. The BCL6 antibody, together with BCL2 antibody, is also a useful aid in classification of mantle cell lymphomas, and nodular, lymphocyte-predominance Hodgkin's lymphoma. BCL6 protein is not expressed in B-CLL, hairy cell leukemia, mantle cell and marginal-zone derived lymphomas
Catalog No. 50-238-5776
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.