Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCL6B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103502
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103502 Supplier Invitrogen™ Supplier No. PA5103502
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BCL6B.

BCL6B acts as a sequence-specific transcriptional repressor in association with BCL6. BCL6B may function in a narrow stage or be related to some events in the early B-cell development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCL6B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BCL6B
Gene Accession No. O88282, Q8N143
Gene Alias B cell CLL/lymphoma 6, member B; B cell CLL/lymphoma 6B; BAZF; B-cell CLL/lymphoma 6 member B protein; B-cell CLL/lymphoma 6, member B; B-cell CLL/lymphoma 6, member B (zinc finger protein); B-cell CLL/lymphoma 6B; Bcl6-associated zinc finger protein; Bcl6b; BCL6B transcription repressor; BCL6B, transcription repressor; LOW QUALITY PROTEIN: B-cell CLL/lymphoma 6 member B protein; RGD1563179; ZBTB28; zinc finger protein 62; ZNF62
Gene Symbols BCL6B
Host Species Rabbit
Immunogen A synthesized peptide derived from human BCL6B(Accession Q8N143), corresponding to amino acid residues L145-K195.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12029, 255877
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.