Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL7B Monoclonal Antibody (OTI2D9), TrueMAB™, OriGene
SDP

Catalog No. 501682826
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2826 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2826 Supplier OriGene Supplier No. TA809485
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive regulator of apoptosis. Plays a role in the Wnt signaling pathway, negatively regulating the expression of Wnt signaling components CTNNB1 and HMGA1. Involved in cell cycle progression, maintece of the nuclear structure and stem cell differentiation. May play a role in lung tumor development or progression.

Specifications

Antigen BCL7B
Applications Western Blot
Classification Monoclonal
Clone OTI2D9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene BCL7B
Gene Accession No. Q9BQE9
Gene Alias BCL tumor suppressor 7B
Gene Symbols BCL7B
Host Species Mouse
Immunogen Full length human recombit protein of human BCL7B produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9275
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.