Learn More
ACROBiosystems Recombinant Protein;Human Bcl-x / BCL2L1 Protein, GST Tag;E.coli;100UG;BC1,BCL2L1,Bcl-XL,Bcl2-L-1,BCL-XL/S,BCL2L,BCLX,BCLXL,BCLXS,Bcl-X,bcl-xS
Supplier: ACROBiosystems BC1H513A100UG
Recombinant Protein;Human Bcl-x / BCL2L1 Protein, GST Tag;100UG;Human Bcl-x, GST Tag (BC1-H513a) is expressed from E.coli cells. It contains AA Met 1 - Arg 212 (Accession # AAH19307).;This protein carries a GST tag at the N-terminus. The protein has a calculated MW of 50.5 kDa. The protein migrates as 46 kDa under reducing (R) condition (SDS-PAGE).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.