Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCMA/TNFRSF17 Antibody (belantamab), R&D Systems™
SDP

Catalog No. NB465765
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB465765 100 μg
NB465763 50 μg
NB465764 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB465765 Supplier R&D Systems Supplier No. NBP328602100UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Human Monoclonal Antibody

BCMA/TNFRSF17 Monoclonal specifically detects BCMA/TNFRSF17 in Human samples. It is validated for ELISA∣Flow Cytometry∣Functional.

Specifications

Antigen BCMA/TNFRSF17
Applications ELISA, Flow Cytometry, Functional Assay
Classification Monoclonal
Clone belantamab
Conjugate Unconjugated
Formulation Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Gene Accession No. Q02223
Gene Alias B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, tumor necrosis factor receptor superfamily, member 17
Host Species Human
Immunogen TNFRSF17/ BCMA/ CD269
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 608
Reconstitution Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Target Species Human
Content And Storage Store at -20°C in powder form. Store at -80°C once reconstituted.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.