Description
- Specialized coating to reduce particles, fibers and micro-organisms.
- Smooth surface reduces surface particle contamination.
- Retains toner from copiers and laser printers to reduce smudging.
- Full lot tracing on every case.
- Double packed for cleanliness.
Specifications
Specifications
| Material | Bond Paper |
| Ruled | No |
| Color | Blue |
| Cover Color | Blue |
| Autoclavable | Not Autoclavable |
| Disposable | Single-use |
| Product Type | Cleanroom Bond |
| Sterility | Non-sterile |