Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD A431 + CAL A/OKA ACID (2HR) Lysate

Catalog No. BDB612591 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612591 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612591 Supplier BD Supplier No. 612591

missing translation for 'validMainframeMsg'

Includes

Human Epidermal Carcinoma

Provided in a denatured format allowing for immediate loading onto an SDS-PAGE gel

  • Control for western blotting application
  • Source rich in endogenously expressed proteins

TRUSTED_SUSTAINABILITY

Specifications

Tissue Epidermal Carcinoma
Preparation Method A431 Cells were grown to 80 to 90% confluency, Serum starved overnight, then treated with 10nM Calyculin A and 500nM Okadaic Acid for 2 Hours at 37°C
Quantity 500 μg
Storage Buffer 1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol
Host Species Human
Concentration 1 mg/mL
Content And Storage -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.