Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD A431 + EGF Cell Lysate

Catalog No. BDB611448 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611448 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611448 Supplier BD Supplier No. 611448

missing translation for 'validMainframeMsg'

Includes

Human Epidermoid Carcinoma

Cell or tissue lysates are a useful control for the western blotting application

A source that can be rich in endogenously expressed proteins, this lysate is convenient to use and provided in a denatured format allowing for immediate loading onto an SDS-PAGE gel. A431 cells (Human epithelial carcinoma; ATCC CRL-1555) were grown to 80-90% confluency, serum starved overnight, then treated with 100ng/mL EGF for 5 minutes at 37°C.

  • Storage buffer: SDS-PAGE buffer (62mM Tris pH 6.8, 2% SDS, 0.9% b-mercaptoethanol, 0.003% bromophenol blue, 5% glycerol)

Western blot

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot
Tissue Epidermoid Carcinoma
Quantity 500 μg
Storage Buffer 1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol
Host Species Human
Concentration 1 mg/mL
Content And Storage -20°C

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.