Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Ac-DEVD-CHO Inhibitor

Catalog No. BDB556465 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556465 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556465 Supplier BD Supplier No. 556465

Requires

Apoptotic and non-apoptotic cells, phosphate buffered saline (PBS) Wash Buffer: 140mM NaCl, 2.7mM KCl, 10mM KH2PO4 dissolved in distilled, autoclaved water. Adjust the pH to 7.4 using hydrochloric acid, Cell Lysis Buffer: 10mM Tris (pH 7.5), 130mM NaCl, 1% Triton -X-100, 10mM NaPi, 10mM NaPPi, Caspase-3 (CPP32), Ac-DEVD-CHO Inhibitor, and substrate Ac-DEVD-AMC, Protease Assay Buffer: 20mM HEPES (pH 7.5), 10% glycerol, 2mM DTT

To identify and quantify Caspase-3 activity in apoptotic cells, and to study events downstream of Caspase-3 activation

Members of ICE/CED-3 cysteine protease family have key roles in inflammation and mammalian apoptosis. The ICE family member Caspase-3 (also known as CPP32, Yama, apopain) is activated early in apoptosis and appears to be involved in proteolysis of several important molecules, including poly (ADP ribose) polymerase (PARP)

  • Synthetic tetrapeptide inhibitor for Caspase-3
  • Used in protease assays
  • Cleaves PARP from 116kD to 85kD residual fragment
  • Cleavage site in PARP is C-terminal to Asp-216
  • Upstream sequence of cleavage site, DEVD (Asp-Glu-Val-Asp), is utilized as basis for highly specific Caspase-3 substrate, Ac-DEVD-AMC and Caspase-3 inhibitor,Ac-DEVD-CHO
  • Storage Buffer: Lyophilized powder
  • Format: Purified

Functional assay

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Functional assay
Storage Buffer Lyophilized powder
Content And Storage -20°C
Quantity 1 mg
Molecular Weight (g/mol) 502.5
Inhibitors Ac-DEVD-CHO
Form Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.