Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Annexin V Binding Buffer

Catalog No. BDB556454 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556454 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556454 Supplier BD Supplier No. 556454
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Formulated for use in flow cytometric assays utilizing Annexin V

  • Composed of 0.2μm sterile filtered 0.1M Hepes (pH 7.4), 1.4M NaCl, and 25mM CaCl2 solution
  • 1X working solution should be made by diluting the 10X concentrate
  • Remaining 1X solution at the end of the experiment should be discarded
  • Format: Buffer

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mL
Content And Storage Store at 4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.