Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Annexin V-FITC
Click to view available options
Quantity:
100 Tests
200 Tests
25 Tests
Description
Identifies apoptosis at an earlier stage than assays based on nuclear changes such as DNA fragmentation.
- Format: FITC
Flow Cytometry

Specifications
Specifications
Quantity | 25 Tests |
Sample Volume | 5 μL |
Storage Buffer | Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
Content And Storage | 4°C |
For Use With (Application) | Flow Cytometry |
Conjugate | FITC |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction