Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Annexin V-PE

Catalog No. BDB560930 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
100 Tests
200 Tests
25 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560930 25 Tests
BDB556422 100 Tests
BDB556421 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BDB560930 Supplier BD Supplier No. 560930
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Serves as a sensitive probe for flow cytometric analysis of cells that are undergoing apoptosis

  • Format: PE

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 Tests
Sample Volume 5 μL
Storage Buffer Aqueous Buffered solution containing BSA and ≤0.09% Sodium Azide
Content And Storage 4°C
For Use With (Application) Flow Cytometry

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.