Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Annexin V: PE Apoptosis Detection Kit I

Catalog No. bdb559763 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559763 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559763 Supplier BD Supplier No. 559763
Only null left
Add to Cart
Add to Cart

Includes

Annexin V-PE, 7-AAD, and Annexin V Binding Buffer

Contains apoptotic program characterized by certain morphologic features, including loss of plasma membrane asymmetry and attachment, condensation of the cytoplasm and nucleus, and internucleosomal cleavage of DNA

  • Process involves staining which precedes the loss of membrane integrity accompanying the latest stages of cell death resulting from either apoptotic or necrotic processes

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit
Content And Storage 4°C
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.