Learn More
Description
Includes
FITC-dUTP, PI/RNase Staining Buffer, Reaction Buffer, Rinsing Buffer, Wash Buffer, TdT Enzyme, Negative Control Cells, Positive Control Cells
- Reagents are provided to process 50 tests
Flow cytometry
Specifications
Specifications
| Quantity | 1 Kit |
| For Use With (Application) | Flow Cytometry |
| Form | Liquid |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.