Learn More
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | M Broth |
| Product Type | Microbiology Media |
| Formulation | Yeast extract 5g/L, Pancreatic digest of Casein 12.5g/L, D-Mannose 2g/L, Sodium Citrate 5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 5g/L, Manganese Chloride 0.14g/L, Magnesium Sulfate 0.8g/L, Ferrous Sulfate 0.04g/L, Polysorbate 80 0.75g/L |
| For Use With (Application) | Used for cultivating Salmonella in foods and feeds by the accelerated enrichment serology (ES) procedure |
| Packaging Type | Pail |
| Quantity | 2000 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.