Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: AK Agar No.2 (Sporulating Agar)

Catalog No. L10912 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L10912 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L10912 Supplier BD Supplier No. 210912
Add to Cart
Add to Cart
This item is not returnable. View return policy

For isolation, detection and cultivation microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Form Dehydrated
Media Type Arret and Kirshbaum Agar No. 2
Product Type Agar
Formulation Pancreatic digest of Gelatin 6g/L, Pancreatic digest of Casein 4g/L, Yeast extract 3g/L, Beef extract 1.5g/L, Dextrose 1g/L, Agar 15g/L, Manganous Sulfate 0.3g/L
For Use With (Application) For preparation of spore suspensions for use in procedures for the detection of antibiotic residues in Milk and dairy products
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.