Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Brain Heart Infusion Broth, Modified

Catalog No. b99070 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Form:
Dehydrated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
B99070 Dehydrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B99070 Supplier BD Supplier No. 299070
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

For cultivation of fastidious and nonfastidious microorganisms, including aerobic and anaerobic bacteria from variety of clinical and nonclinical materials

  • Formulation: Brain heart infusion from (solids) 3.5g/L, Peptic digest of Animal Tissue 15g/L, Pancreatic digest of Casein 10g/L, Dextrose 2g/L, Sodium Chloride 5g/L, Disodium Phosphate 2.5g/L

TRUSTED_SUSTAINABILITY

Specifications

Form Dehydrated
Media Type Brain Heart Infusion Broth
Product Type Microbiology Media
Formulation Brain heart infusion from (solids) 3.5g/L, Peptic digest of Animal Tissue 15g/L, Pancreatic digest of Casein 10g/L, Dextrose 2g/L, Sodium Chloride 5g/L, Disodium Phosphate 2.5g/L
For Use With (Application) Cultivation of fastidious and non-fastidious microorganisms
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.