Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Brucella Agar

Catalog No. B11086 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11086 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11086 Supplier BD Supplier No. 211086
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dehydrated form for the convenience of the laboratorian and bioprocessing specialist

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CCAM, ISO, USDA
Form Dehydrated
Media Type Brucella Agar
Product Type Agar
Formulation Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Dextrose 1g/L, Yeast extract 2g/L, Sodium Chloride 5g/L, Sodium Bisulfite 0.1g/L, Agar 15g/L
For Use With (Application) Cultivation of Brucella organisms
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.