Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Columbia CNA Agar

Catalog No. B12104 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
Packaging Type:
Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
B12104 500 g Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B12104 Supplier BD Supplier No. 212104
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For selective isolation of gram positive cocci

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Columbia CNA Agar
Product Type Agar
Formulation Pancreatic digest of Casein 12g/L, Peptic digest of Animal Tissue 5g/L, Yeast extract 3g/L, Beef extract 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Colistin 10mg/L, Nalidixic Acid 10mg/L
For Use With (Application) For selective isolation of gram positive cocci
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.