Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Corn Meal Agar

Catalog No. B11132 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Content And Storage:
2°C to 8°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
B11132 2°C to 8°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B11132 Supplier BD Supplier No. 211132
Add to Cart
Add to Cart
This item is not returnable. View return policy

For cultivation of fungi, with incorporation of Polysorbate 80, utilized for testing of Candida species for their ability to produce chlamydospores

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Corn Meal Agar
Product Type Agar
Formulation Corn Meal infusion from (solids) 2g/L, Agar 15g/L
For Use With (Application) General purpose medium for the cultivation of fungi, With the incorporation of Polysorbate 80, utilized for the testing of Candida species for their ability to produce chlamydospores
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.